Corning™ Microvolume Pipet Tips, Nonsterile
Provide accurate, reliable performance in the stated volume range
Supplier:  Corning™ 4826
 View more versions of this product
Catalog No. 07-200-522
Description
Description
  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
Specifications
0.1 to 10 μL
Natural
Rack
Rack
Polypropylene
Non-sterile
DNase, RNase, and Pyrogen free
Microvolume, Round
Gilson™ Pipetman™, Universal Fit Ultramicro