missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Microvolume Pipet Tips, Nonsterile
Provide accurate, reliable performance in the stated volume range
$178.41 - $1081.69
Specifications
Sterility | Non-sterile |
---|---|
Color | Natural |
Clean Claims | DNase, RNase, and Pyrogen free |
Tip Style | Microvolume, Round |
Material | Polypropylene |
Catalog Number | Mfr. No. | Volume (Metric) | Format | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Format | Price | Quantity | ||||
07-200-525
|
Corning™
4834 |
0.5 to 10 μL | Rack |
Case of 960 Each for $183.27
|
|
||||
07-200-280
|
Corning™
C4840 |
0.2 to 10 μL | Bulk |
Case of 10000 Each for $1,081.69
|
|
||||
07-200-526
|
Corning™
4901 |
0.5 to 10 μL | Bulk |
Case of 10000 Each for $1,055.84
|
|
||||
07-200-522
|
Corning™
4826 |
0.1 to 10 μL | Rack |
Case of 960 Each for $178.41
|
|
||||
Description
- Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
Non-sterile | |
DNase, RNase, and Pyrogen free | |
Polypropylene |
Natural | |
Microvolume, Round |