Minigrip™ Lab Guard™ Specimen Bags
Protect from inadvertent exposure to leakage and contamination
Supplier:  Minigrip™ SBL2X69S
 View more versions of this product
Catalog No. 01823
Description
Description
Bags with the destroyable biohazard symbol feature allow uncontaminated bag remains to be disposed in regular waste; if bag remains are contaminated, dispose in RED waste.
  • Zippered polyethylene three-wall bags with attached document compartment
  • Sample and paperwork stay together to reduce clerical errors and misidentification
  • TearZone™ EZ open feature means no leaks
  • Available with convenient DispenserBag™ and DispenserBox packaging
Order Info
Available plain or with orange and black biohazard labeling.†
Compliance
Comply with OSHA guidelines. STAT Specimen Transport Bags are FDA registered.
†Features a bright red requisition pocket and STAT labeling to easily identify urgent tests.
Specifications
Specifications
STAT Specimen Transport Reclosable Bag with bold STAT, biohazard symbol and clear back
For transportation of specimens
1000/Cs.
9 x 6 in. (23 x 15cm)
23 cm
15 cm
Zip Seal
Polyethylene
1.75 mil
9 in.
6 in.