missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minigrip™ Lab Guard™ Specimen Bags
Protect from inadvertent exposure to leakage and contamination
$140.00 - $787.71
Specifications
Closure Type | Zip Seal |
---|---|
For Use With (Application) | For transportation of specimens |
Material | Polyethylene |
Catalog Number | Mfr. No. | Dimensions (L x W) | Description | Quantity | Thickness (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Description | Quantity | Thickness (English) | Price | Quantity | ||
22-310302
|
Minigrip™
SBL2X1215B |
15 x 12 in. (38.1 x 30.5cm) | 3-Walled Biohazard Specimen Bag | 1000/Cs. | 2 mil |
Case of 1000 Each for $787.71
|
|
||
01823
|
Minigrip™
SBL2X69S |
9 x 6 in. (23 x 15cm) | STAT Specimen Transport Reclosable Bag with bold STAT, biohazard symbol and clear back | 1000/Cs. | 1.75 mil |
Case of 1000 Each for $250.03
|
|
||
22-306-325
|
Minigrip™
SBL2X46B |
6 x 4 in. (15.2 x 10.1cm) | 3-Walled Biohazard Specimen Bag | 1000/Cs. | 1.75 mil |
Case of 1000 Each for $140.00
|
|
||
Description
Bags with the destroyable biohazard symbol feature allow uncontaminated bag remains to be disposed in regular waste; if bag remains are contaminated, dispose in RED waste.
- Zippered polyethylene three-wall bags with attached document compartment
- Sample and paperwork stay together to reduce clerical errors and misidentification
- TearZone™ EZ open feature means no leaks
- Available with convenient DispenserBag™ and DispenserBox packaging
Order Info
Available plain or with orange and black biohazard labeling.†
Compliance
Comply with OSHA guidelines. STAT Specimen Transport Bags are FDA registered.
†Features a bright red requisition pocket and STAT labeling to easily identify urgent tests.
Specifications
Zip Seal | |
For transportation of specimens | |
Polyethylene |