missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minigrip™ Lab Guard™ Specimen Bags with TearZone™ and Destroyable Biohazard Symbol
Safe and durable specimen-handling reclosable bags feature “open and close” zipper technology and grip edges
Supplier: Minigrip™ MGTZ69BLU
Description
With TearZone™ destroyable biohazard symbol feature.
- Convenient 6 by 9 in. size
- Color codes can be used to identify a specific urgency or department
- TearZone™ feature will not leak — contains leak to one bag for overall cleanup and savings
- Tears open easily at processing — no need for sharps to open the bag quickly
- Grip edges make opening easier with gloved hands
- Vivid biohazard risk warning graphic
- Separate pouch for paperwork storage
- Option to dispose unsoiled bags in regular waste means additional cost saving
Specifications
Biohazard Specimen Transport Bag | |
Biohazard specimen transport | |
Virgin Polyethylene | |
2 mil | |
9 in. | |
6 in. |
Blue | |
TearZone™ and destroyable biohazard symbol | |
0.279 mm | |
9 x 6 in. (22.8 x 15.2cm) | |
22.8 cm | |
15.2 cm |