missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minigrip™ Lab Guard™ Specimen Bags with TearZone™ and Destroyable Biohazard Symbol
Safe and durable specimen-handling reclosable bags feature “open and close” zipper technology and grip edges
$136.07 - $246.50
Specifications
Product Type | Biohazard Specimen Transport Bag |
---|---|
For Use With (Application) | Biohazard specimen transport |
Includes | TearZone™ and destroyable biohazard symbol |
Material | Virgin Polyethylene |
Thickness (Metric) | 0.279 mm |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
22-311-105
|
Minigrip™
MGTZ69BLU |
Blue |
Case of 1000 Each for $229.15
|
|
|||||
22-311-107
|
Minigrip™
MGTZ69RD |
Red |
Case of 1000 Each for $244.18
|
|
|||||
22-311-108
|
Minigrip™
MGTZ69S |
Red Tint |
Case of 1000 Each for $246.50
|
|
|||||
22311109
|
Minigrip™
MGTZ69YEL |
Yellow |
Case of 1000 Each for $136.07
|
|
|||||
22-311--111
|
Minigrip™
MGTZ69PUR |
Purple |
Case of 1000 Each for $244.12
|
|
|||||
Description
With TearZone™ destroyable biohazard symbol feature.
- Convenient 6 by 9 in. size
- Color codes can be used to identify a specific urgency or department
- TearZone™ feature will not leak — contains leak to one bag for overall cleanup and savings
- Tears open easily at processing — no need for sharps to open the bag quickly
- Grip edges make opening easier with gloved hands
- Vivid biohazard risk warning graphic
- Separate pouch for paperwork storage
- Option to dispose unsoiled bags in regular waste means additional cost saving
Specifications
Biohazard Specimen Transport Bag | |
TearZone™ and destroyable biohazard symbol | |
0.279 mm | |
9 x 6 in. (22.8 x 15.2cm) | |
22.8 cm | |
15.2 cm |
Biohazard specimen transport | |
Virgin Polyethylene | |
2 mil | |
9 in. | |
6 in. |