Minigrip™ SPECI-GARD™ Specimen Biohazard Transport Bags
Used for specimen transport in hospitals and labs
Supplier:  Minigrip™ UF95600BIO
 View more versions of this product
Catalog No. 22-314-257
Explore available promotions
Description
Description
  • Features liquid-tight, Press and Close™ adhesive closure system — simply peel away the release liner and press to close
  • With printed biohazard logo
  • Separate outside pocket holds the requisition slip
  • Traditional scored opening
  • No glove-to-adhesive contact, ensuring ease of use
  • Once closed, the bag stays closed, even in pneumatic tube systems
  • Remains closed if air is trapped inside, or if “grabbed,” will not “pop open”
  • Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging
  • To open, simply snap at score
  • Disposable, eliminating the possibility of cross-contamination caused by reusing bags
  • Manufactured with Micro-Tex™ polyethylene for slip-free handling
Order Info
Cat. No. 22-310-001 contains an additional pre-inserted super absorbent tested to absorb up to 60mL of liquid.
Specifications
Specifications
Biohazard Specimen Transport Bag
Single-use
Press and Close™ liquid tight adhesive closure, 2 pockets, biohazard label
10 in.
6 in.
10 in.
Clear
Used for specimen transport in hospitals and labs
Polyethylene
25.4 cm
15.24 cm
25.4 cm