missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minigrip™ SPECI-GARD™ Specimen Biohazard Transport Bags
Used for specimen transport in hospitals and labs
$468.29 - $893.24
Specifications
Product Type | Biohazard Specimen Transport Bag |
---|---|
Color | Clear |
Disposable | Single-use |
For Use With (Application) | Used for specimen transport in hospitals and labs |
Material | Polyethylene |
Catalog Number | Mfr. No. | Includes | Length (English) | Width (English) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Length (English) | Width (English) | Price | Quantity | ||||
22-314-257
|
Minigrip™
UF95600BIO |
Press and Close™ liquid tight adhesive closure, 2 pockets, biohazard label | 10 in. | 6 in. |
Case of 1000 Each for $468.29
|
|
||||
22-310-094
|
N/A | Press and Close™ liquid tight adhesive closure, single pocket (2 wall), biohazard label | 6 in. | 6 in. |
Case of 2000 Each for $893.24
|
|
||||
22-310-001
|
N/A | Double pockets, with red and black print, biohazard label | 10 in. | 6 in. |
Case of 1000 Each for $700.48
|
|
||||
22-310-004
|
N/A | Press and Close™ liquid tight adhesive closure and instructions, biohazard label | 10 in. | 6 in. |
Case of 1000 Each for $617.48
|
|
||||
Description
- Features liquid-tight, Press and Close™ adhesive closure system — simply peel away the release liner and press to close
- With printed biohazard logo
- Separate outside pocket holds the requisition slip
- Traditional scored opening
- No glove-to-adhesive contact, ensuring ease of use
- Once closed, the bag stays closed, even in pneumatic tube systems
- Remains closed if air is trapped inside, or if “grabbed,” will not “pop open”
- Available in single-pocket versions when requisitions are not used, or in diagnostic kit packaging
- To open, simply snap at score
- Disposable, eliminating the possibility of cross-contamination caused by reusing bags
- Manufactured with Micro-Tex™ polyethylene for slip-free handling
Order Info
Cat. No. 22-310-001 contains an additional pre-inserted super absorbent tested to absorb up to 60mL of liquid.
Specifications
Biohazard Specimen Transport Bag | |
Single-use | |
Polyethylene |
Clear | |
Used for specimen transport in hospitals and labs |