Minigrip™ SPECI-GARD™ STAT Specimen Transport Bag
Easily identifies specimens for priority handling; ideal for pneumatic tube systems
Supplier:  Minigrip™ UF95600STBR
 View more versions of this product
Catalog No. 22-307-949
Explore available promotions
Description
Description
  • Double pocket, three-wall bag features Press-and Close™ one-touch safety seal for specimens and integral outside requisition pocket
  • Prominent “STAT” copy displayed on bag with biohazard symbol
  • Seals in specimens to reduce the risk of contact with dangerous body fluids
  • Not reusable; once sealed, cannot be reopened without compromising the integrity of the bag
Order Info
Dimensions: 6W x 10 in. H. Case of 1000 in packs of 100.
Compliance
Meets OSHA and NCCLS requirements for liquid-tight transport
Specifications
Specifications
Specimen Transport Bag
Single-use
Double pockets bag with Press and Close™ liquid tight closure, transparent red bag printed with biohazard logo and instructions for use
15.24 cm
25.4 cm
Red
For specimen transport
6 in.
10 in.