missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Minigrip™ SPECI-GARD™ STAT Specimen Transport Bag
Easily identifies specimens for priority handling; ideal for pneumatic tube systems
$426.91 - $426.91
Specifications
Product Type | Specimen Transport Bag |
---|---|
Disposable | Single-use |
For Use With (Application) | For specimen transport |
Width (English) | 6 in. |
Width (Metric) | 15.24 cm |
Catalog Number | Mfr. No. | Color | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Includes | Price | Quantity | ||||
22-307-949
|
Minigrip™
UF95600STBR |
Red | Double pockets bag with Press and Close™ liquid tight closure, transparent red bag printed with biohazard logo and instructions for use |
Case of 1000 Each for $426.91
|
|
||||
Description
- Double pocket, three-wall bag features Press-and Close™ one-touch safety seal for specimens and integral outside requisition pocket
- Prominent “STAT” copy displayed on bag with biohazard symbol
- Seals in specimens to reduce the risk of contact with dangerous body fluids
- Not reusable; once sealed, cannot be reopened without compromising the integrity of the bag
Order Info
Dimensions: 6W x 10 in. H. Case of 1000 in packs of 100.
Compliance
Meets OSHA and NCCLS requirements for liquid-tight transport
Specifications
Specimen Transport Bag | |
For specimen transport | |
15.24 cm | |
25.4 cm |
Single-use | |
6 in. | |
10 in. |