missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Minoxidil, Thermo Scientific Chemicals

Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels.

Supplier:  Thermo Scientific Chemicals J6180314

 View more versions of this product

Catalog No. AAJ6180314


Explore available promotions

Description

Description

Minoxidil is used as antihypertensive, antialopecia agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
  • Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure

Specifications

Specifications

Minoxidil
248°C to 265°C
C9H15N5O
25 g
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
4201
Crystalline Powder
38304-91-5
White
MFCD00063409
14,6203
Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol.
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
209.253
209.25

Chemical Identifiers

38304-91-5
209.253
ZIMGGGWCDYVHOY-UHFFFAOYSA-N
4201
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
C9H15N5O
MFCD00063409
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 253-874-2

RTECSNumber : UV8200000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only