Learn More
Minoxidil, Thermo Scientific Chemicals
Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels.
$56.78 - $367.07
Chemical Identifiers
CAS | 38304-91-5 |
---|---|
Molecular Formula | C9H15N5O |
Molecular Weight (g/mol) | 209.253 |
MDL Number | MFCD00063409 |
InChI Key | ZIMGGGWCDYVHOY-UHFFFAOYSA-N |
Synonym | minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox |
PubChem CID | 4201 |
IUPAC Name | 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine |
SMILES | C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ6180303
|
Thermo Scientific Chemicals
J6180303 |
1 g |
Each for $56.78
|
|
||||||
AAJ6180306
|
Thermo Scientific Chemicals
J6180306 |
5 g |
Each for $204.53
|
|
||||||
AAJ6180314
|
Thermo Scientific Chemicals
J6180314 |
25 g |
Each for $367.07
|
|
||||||
Description
Minoxidil is used as antihypertensive, antialopecia agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
- Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure
Chemical Identifiers
38304-91-5 | |
209.253 | |
ZIMGGGWCDYVHOY-UHFFFAOYSA-N | |
4201 | |
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O |
C9H15N5O | |
MFCD00063409 | |
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox | |
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine |
Specifications
38304-91-5 | |
White | |
MFCD00063409 | |
14,6203 | |
Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol. | |
C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O | |
209.253 | |
209.25 | |
Minoxidil |
248°C to 265°C | |
C9H15N5O | |
1 g | |
minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox | |
ZIMGGGWCDYVHOY-UHFFFAOYSA-N | |
3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine | |
4201 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 253-874-2
RTECSNumber : UV8200000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only