missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mirus Bio™ Ingenio™ Electroporation Kit

Use the Ingenio™ Electroporation Kit for high efficiency electroporation using any instrument.

Supplier:  Mirus Bio™ MIR50115

 View more versions of this product

Catalog No. MIR50115


Explore available promotions

Description

Description

  • High efficiency electroporation of hard to transfect cell lines and primary cells
  • Compatible with most conventional electroporation devices including Lonza-Amaxa™, Bio-Rad™ or Harvard BTX™
  • Ideal for plasmid or siRNA electroporation

  • Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
  • Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
  • Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
  • High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Specifications

Specifications

Plasmid or siRNA electroporation
Electroporation Kit
50
4°C
Lonza Amaxa™ Nucleofector™ electroporator
1 x 12.5mL Ingenio solution, 50 of 0.2cm cuvettes, 50 cell droppers
Sterile
Exponential Decay Wave
Videos
SDS
Documents

Documents

Promotions

Promotions