missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mirus Bio™ Ingenio™ Electroporation Kit
Use the Ingenio™ Electroporation Kit for high efficiency electroporation using any instrument.
$886.55 - $2879.64
Specifications
For Use With (Application) | Plasmid or siRNA electroporation |
---|---|
Product Type | Electroporation Kit |
Sterility | Sterile |
Content And Storage | 4°C |
Pulse Form | Exponential Decay Wave |
Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Quantity | Price | Quantity | |||
MIR50112
|
Mirus Bio™
MIR50112 |
Lonza Amaxa™ Nucleofector™ electroporator | 1 x 6.25mL Ingenio solution, 25 of 0.2cm cuvettes, 25 cell droppers | 25 |
Each for $1,493.74
|
|
|||
MIR50116
|
Mirus Bio™
MIR50116 |
Bio-Rad™ gene pulser and Harvard-BTX™ electroporator | 1 x 12.5mL Ingenio solution, 50 of 0.4cm cuvettes, 50 cell droppers | 50 |
Each for $1,549.83
|
|
|||
MIR50115
|
Mirus Bio™
MIR50115 |
Lonza Amaxa™ Nucleofector™ electroporator | 1 x 12.5mL Ingenio solution, 50 of 0.2cm cuvettes, 50 cell droppers | 50 |
Each for $1,549.83
|
|
|||
MIR50113
|
Mirus Bio™
MIR50113 |
Bio-Rad™ gene pulser and Harvard-BTX™ electroporator | 1 x 6.25mL Ingenio solution, 25 of 0.4cm cuvettes, 25 cell droppers | 25 |
Each for $886.55
|
|
|||
MIR50118
|
Mirus Bio™
MIR50118 |
Lonza Amaxa™ Nucleofector™ electroporator | 1 x 12.5mL Ingenio solution, 100 of 0.2cm cuvettes, 100 cell droppers | 100 |
Each for $2,879.64
|
|
|||
MIR50119
|
Mirus Bio™
MIR50119 |
Bio-Rad™ gene pulser and Harvard-BTX™ electroporator | 2 x 12.5mL Ingenio solution, 100 of 0.4cm cuvettes, 100 cell droppers | 100 |
Pack of 100 Each for $2,879.64
|
|
|||
Description
- High efficiency electroporation of hard to transfect cell lines and primary cells
- Compatible with most conventional electroporation devices including Lonza-Amaxa™, Bio-Rad™ or Harvard BTX™
- Ideal for plasmid or siRNA electroporation
- Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
- Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Specifications
Plasmid or siRNA electroporation | |
Sterile | |
Exponential Decay Wave |
Electroporation Kit | |
4°C |