missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mirus Bio™ Ingenio™ Electroporation Kit
Use the Ingenio™ Electroporation Kit for high efficiency electroporation using any instrument.
Supplier: Mirus Bio™ MIR50118
Description
- High efficiency electroporation of hard to transfect cell lines and primary cells
- Compatible with most conventional electroporation devices including Lonza-Amaxa™, Bio-Rad™ or Harvard BTX™
- Ideal for plasmid or siRNA electroporation
- Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
- Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Specifications
Plasmid or siRNA electroporation | |
Electroporation Kit | |
100 | |
4°C |
Lonza Amaxa™ Nucleofector™ electroporator | |
1 x 12.5mL Ingenio solution, 100 of 0.2cm cuvettes, 100 cell droppers | |
Sterile | |
Exponential Decay Wave |