Moldex™ 1500 Series N95 Healthcare Particulate Respirators and Surgical Masks
N95 healthcare and surgical mask feature ASTM Level 3 protection (BFE >99.9%) and are ideal for use in healthcare settings and hospitals.
Supplier:  Moldex™ MDX1510
 View more versions of this product
Catalog No. 29-636-0706
Explore available promotions
Description
Description
  • Molded nose bridge with soft foam nose cushion provides easy, comfortable, dependable fit
  • Dura-Mesh™ shell prevents filter media contact by contaminated gloves or hands; resists collapsing, even in heat and humidity
  • Nonlatex straps don't irritate or require prestretching
  • Highest ASTM™ level of fluid resistance on the market (160mmHg)
  • Bacterial filter efficiency >99.9%
  • Softspun™ inner lining is soft to the touch
  • Available in four color-coded sizes
  • ASTM Level 3
Order Info
20 respirators per pack.
Specifications
Specifications
N - Not Resistant to Oil
ASTM F1862
Cup/Molded
Surgical N95
Healthcare Particulate Respirator and Surgical Mask
No
Yes
1510
Molded Nose Bridge
Fluid 160 mmHg
Dual Elastic Straps
ANSI/ISEA 110-2009, Section 7.11.1
NIOSH 42 CFR 84, N95
Single-use
Dura-Mesh
X-Small
95%
Softspun™
Soft Foam
20/Box; 8 Boxes/Cs.
Non-sterile
Yes