Learn More
Molybdic acid, MoO3 85% min, Thermo Scientific Chemicals
$75.78 - $935.00
Chemical Identifiers
CAS | 7782-91-4 |
---|---|
Molecular Formula | H2MoO4 |
Molecular Weight (g/mol) | 161.96 |
MDL Number | MFCD00036287 |
InChI Key | VLAPMBHFAWRUQP-UHFFFAOYSA-L |
Synonym | molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 |
PubChem CID | 82208 |
ChEBI | CHEBI:25371 |
IUPAC Name | dihydroxy(dioxo)molybdenum |
SMILES | O[Mo](O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1053722
|
Thermo Scientific Chemicals
A1053722 |
100 g |
Each for $75.78
|
|
||||||
AAA1053736
|
Thermo Scientific Chemicals
A1053736 |
500 g |
Each for $250.19
|
|
||||||
AAA105370E
|
Thermo Scientific Chemicals
A105370E |
2500 g |
Each for $935.00
|
|
||||||
Description
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMolybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.
Solubility
Soluble in dilute ammonium hydroxide, alkali and ammonia. Slightly soluble in water.
Notes
Incompatible with strong bases and strong oxidizing agents.
Chemical Identifiers
7782-91-4 | |
161.96 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
82208 | |
dihydroxy(dioxo)molybdenum |
H2MoO4 | |
MFCD00036287 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
CHEBI:25371 | |
O[Mo](O)(=O)=O |
Specifications
7782-91-4 | |
H2MoO4 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
dihydroxy(dioxo)molybdenum | |
82208 | |
MoO3 85% min | |
Molybdic acid |
100 g | |
MFCD00036287 | |
Soluble in dilute ammonium hydroxide,alkali and ammonia. Slightly soluble in water. | |
O[Mo](O)(=O)=O | |
161.96 | |
CHEBI:25371 | |
3.1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
H319-H335-H373
EINECSNumber : 231-970-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only