missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monobasic Sodium Phosphate, Anhydrous, USP, 98-103%, Spectrum™ Chemical
SO187, 7558-80-7, NaH2PO4
Supplier: Spectrum Chemical Mfg Cor SO18712KG
Description
Spectrum™ Chemical Monobasic Sodium Phosphate, Anhydrous, USP is used as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7558-80-7 | |
| 4.1 to 4.5 | |
| H2NaO4P | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| sodium dihydrogen phosphate | |
| 98 to 103% | |
| Poly Pail |
| 1 | |
| 12 kg | |
| MFCD00003527,MFCD00146206 | |
| [Na+].OP(O)([O-])=O | |
| 119.98 | |
| USP |
Chemical Identifiers
| 7558-80-7 | |
| 119.98 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| [Na+].OP(O)([O-])=O |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| sodium dihydrogen phosphate |