missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Monobasic Sodium Phosphate, Anhydrous, USP, 98-103%, Spectrum™ Chemical
SO187, 7558-80-7, NaH2PO4
$1447.32 - $9513.50
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Formula | H2NaO4P |
Molecular Weight (g/mol) | 119.98 |
MDL Number | MFCD00003527,MFCD00146206 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
IUPAC Name | sodium dihydrogen phosphate |
SMILES | [Na+].OP(O)([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18604658
|
Spectrum Chemical Mfg Cor
SO18725KG |
2.5 kg | Poly Bottle |
Each for $1,447.32
|
|
||||
18604659
|
Spectrum Chemical Mfg Cor
SO18712KG |
12 kg | Poly Pail |
Each for $3,913.35
|
|
||||
18604661
|
Spectrum Chemical Mfg Cor
SO18725KGBL |
25 kg | Fiber Drum |
Each for $6,350.68
|
|
||||
18604660
|
Spectrum Chemical Mfg Cor
SO18745KGBL |
45 kg | Fiber Drum |
Each for $9,513.50
|
|
||||
Description
Spectrum™ Chemical Monobasic Sodium Phosphate, Anhydrous, USP is used as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
[Na+].OP(O)([O-])=O |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
sodium dihydrogen phosphate |
Specifications
7558-80-7 | |
4.1 to 4.5 | |
H2NaO4P | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium dihydrogen phosphate | |
98 to 103% | |
Poly Bottle |
1 | |
2.5 kg | |
MFCD00003527,MFCD00146206 | |
[Na+].OP(O)([O-])=O | |
119.98 | |
USP |