missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monobasic Sodium Phosphate, Anhydrous, USP, 98-103%, Spectrum™ Chemical

SO187, 7558-80-7, NaH2PO4

Supplier:  Spectrum Chemical Mfg Cor SO18745KGBL

 View more versions of this product

Catalog No. 18604660

Explore available promotions

Description

Description

Spectrum™ Chemical Monobasic Sodium Phosphate, Anhydrous, USP is used as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7558-80-7
4.1 to 4.5
H2NaO4P
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
sodium dihydrogen phosphate
98 to 103%
Fiber Drum
1
45 kg
MFCD00003527,MFCD00146206
[Na+].OP(O)([O-])=O
119.98
USP

Chemical Identifiers

7558-80-7
119.98
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
[Na+].OP(O)([O-])=O
H2NaO4P
MFCD00003527,MFCD00146206
sodium dihydrogen phosphate
Videos
SDS
Documents

Documents

Promotions

Promotions