missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Monocaprin 98.0+%, TCI America™

Supplier:  TCI America M10721G

 View more versions of this product

Catalog No. M10721G

Explore available promotions

Description

Description

Specifications

Specifications

Monocaprin
53°C
C13H26O4
1 g
LKUNXBRZDFMZOK-UHFFFAOYNA-N
2,3-dihydroxypropyl decanoate
92926
246.35
Crystalline Powder
26402-22-2
White-Yellow
MFCD00056656
Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol
CCCCCCCCCC(=O)OCC(O)CO
246.35
CHEBI:75551
≥98.0% (GC)

Chemical Identifiers

26402-22-2
246.35
LKUNXBRZDFMZOK-UHFFFAOYNA-N
92926
2,3-dihydroxypropyl decanoate
C13H26O4
MFCD00056656
Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol
CHEBI:75551
CCCCCCCCCC(=O)OCC(O)CO
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0669

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions