missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Monocaprin 98.0+%, TCI America™
$200.75 - $545.81
Chemical Identifiers
CAS | 26402-22-2 |
---|---|
Molecular Formula | C13H26O4 |
Molecular Weight (g/mol) | 246.35 |
MDL Number | MFCD00056656 |
InChI Key | LKUNXBRZDFMZOK-UHFFFAOYNA-N |
Synonym | Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol |
PubChem CID | 92926 |
ChEBI | CHEBI:75551 |
IUPAC Name | 2,3-dihydroxypropyl decanoate |
SMILES | CCCCCCCCCC(=O)OCC(O)CO |
Chemical Identifiers
26402-22-2 | |
246.35 | |
LKUNXBRZDFMZOK-UHFFFAOYNA-N | |
92926 | |
2,3-dihydroxypropyl decanoate |
C13H26O4 | |
MFCD00056656 | |
Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol | |
CHEBI:75551 | |
CCCCCCCCCC(=O)OCC(O)CO |
Specifications
26402-22-2 | |
White-Yellow | |
MFCD00056656 | |
Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol | |
CCCCCCCCCC(=O)OCC(O)CO | |
246.35 | |
CHEBI:75551 | |
≥98.0% (GC) | |
Monocaprin |
53°C | |
C13H26O4 | |
1 g | |
LKUNXBRZDFMZOK-UHFFFAOYNA-N | |
2,3-dihydroxypropyl decanoate | |
92926 | |
246.35 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0669
TSCA : Yes