Covidien Monoject™ Angel Wing™ Safety Male Collection Sets
Designed for use with Angel Wing blood collection sets
Supplier:  Covidien 8881225224
 View more versions of this product
Catalog No. 22-225224
Explore available promotions
Description
Description
  • Safely transfer blood specimens from syringes into blood collection tubes and blood culture bottles
  • Recessed multi-sample needle protected by a plastic shield—easy application of blood collection tube
  • Sets feature needle-shielded holders for blood collection
  • Sterile
Cat. No. 22-225-216 Tube Holder is designed for use with evacuated blood collection tubes; 22-225-224 Dome for use with blood culture and blood collection tubes.
Specifications
Specifications
Multi-sample Blood Collection Set
Sterile
Holder and Safety Cap