Covidien Monoject™ Angel Wing™ Safety Male Collection Sets
Designed for use with Angel Wing blood collection sets
$203.87 - $203.87
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
22-225224
|
Covidien
8881225224 |
Multi-sample Blood Collection Set |
Case of 50 Each for $203.87
|
|
|||||
Description
- Safely transfer blood specimens from syringes into blood collection tubes and blood culture bottles
- Recessed multi-sample needle protected by a plastic shield—easy application of blood collection tube
- Sets feature needle-shielded holders for blood collection
- Sterile
Cat. No. 22-225-216 Tube Holder is designed for use with evacuated blood collection tubes; 22-225-224 Dome for use with blood culture and blood collection tubes.
Specifications
Sterile | |
Holder and Safety Cap |