Covidien Monoject™ Coated EDTA Tubes with Lavender Stoppers
Coated tubes used for laboratory procedures
Supplier:  Covidien 8881311743
 View more versions of this product
Catalog No. 22-311743
Explore available promotions
Description
Description
  • Sterile interior
  • For plasma or whole blood
Specifications
Specifications
Blood Collection Tube
Silicone
10 mL
Liquid EDTA (K3) 0.10mL, 15% Solution
Sterile
16 × 100 mm (0.630 × 3.94 in.)
Product Suggestions
Product Suggestions