Variety Glass Variety Glass Mooney Air Vent for Liquids
Heavy-molded flint glass funnels with smooth rims, inside ribs
Supplier:  Variety Glass 6953
 View more versions of this product
Catalog No. 10-381A
Explore available promotions
Description
Description
  • Patented double vents extend up the stem of each funnel and allow liquid to flow while air escapesVents end around funnel bowl in heavy supporting bead
Specifications
Specifications
9cm
125 mL
Flint Glass
Product Suggestions
Product Suggestions