Variety Glass Variety Glass Mooney Air Vent for Liquids
Heavy-molded flint glass funnels with smooth rims, inside ribs
$38.57 - $198.17
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric)Top | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric)Top | Price | Quantity | |||||
10-381B
|
Variety Glass
6954 |
250 mL | 11.5cm |
N/A
|
|
|||||
10-381A
|
Variety Glass
6953 |
125 mL | 9cm |
N/A
|
|
|||||
10-381C
|
Variety Glass
6955 |
500 mL | 14.5cm |
N/A
|
|
|||||
10-381D
|
Variety Glass
6956 |
1000 mL | 17.5cm |
N/A
|
|
|||||
10-381E
|
Variety Glass
6957 |
2000 mL | 21.5cm |
N/A
|
|
|||||
10-381F
|
Variety Glass
6958 |
4000 mL | 27cm |
N/A
|
|
|||||
Description
- Patented double vents extend up the stem of each funnel and allow liquid to flow while air escapesVents end around funnel bowl in heavy supporting bead
Specifications
Flint Glass |