Variety Glass Variety Glass Mooney Air Vent for Liquids
Heavy-molded flint glass funnels with smooth rims, inside ribs
Supplier:  Variety Glass 6956
 View more versions of this product
Catalog No. 10-381D
Explore available promotions
Description
Description
  • Patented double vents extend up the stem of each funnel and allow liquid to flow while air escapesVents end around funnel bowl in heavy supporting bead
Specifications
Specifications
17.5cm
1000 mL
Flint Glass
Product Suggestions
Product Suggestions