missing translation for 'onlineSavingsMsg'
Learn More

Morin hydrate

CAS: 654055-01-3 | C15H10O7 | 302.24 g/mol

Supplier:  Thermo Scientific Chemicals 354000100

 View more versions of this product

Catalog No. AC354000100

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Morin hydrate
300.0°C
Authentic
C15H10O7
10 g
14, 6269
Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water, soluble in organic solvents
OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
302.24
302.24
654055-01-3,480-16-0
Yellow to Brown
Glass bottle
MFCD00217054
18, 239
morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
YXOLAZRVSSWPPT-UHFFFAOYSA-N
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
16219651
Powder

Chemical Identifiers

654055-01-3
302.24
YXOLAZRVSSWPPT-UHFFFAOYSA-N
16219651
OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
C15H10O7
MFCD00217054
morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

Warning

EINECSNumber : 207-542-9

RTECSNumber : LK8749000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only