missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Morin hydrate, Thermo Scientific Chemicals

$73.53 - $131.71

Chemical Identifiers

CAS 654055-01-3
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.24
MDL Number MFCD00217054
InChI Key YXOLAZRVSSWPPT-UHFFFAOYSA-N
Synonym morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
PubChem CID 16219651
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
SMILES OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC354000050
SDS View Documents
Thermo Scientific Chemicals
354000050
5 g Glass bottle
Each for $73.53
 
AC354000100
SDS View Documents
Thermo Scientific Chemicals
354000100
10 g Glass bottle
Each for $131.71
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

654055-01-3
302.24
YXOLAZRVSSWPPT-UHFFFAOYSA-N
16219651
OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
C15H10O7
MFCD00217054
morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate

Specifications

654055-01-3,480-16-0
Yellow to Brown
Glass bottle
MFCD00217054
18, 239
morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
YXOLAZRVSSWPPT-UHFFFAOYSA-N
2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
16219651
Powder
300.0°C
Authentic
C15H10O7
5 g
14, 6269
Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water, soluble in organic solvents
OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1
302.24
302.24
Morin hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

Warning

EINECSNumber : 207-542-9

RTECSNumber : LK8749000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only