Morphix Technologies ChromAir™ Passive Sampling Badges
Built-in scale features six dose ranges. Morphix Technologies ChromAir Passive Sampling Quantitative Badges provide a cost-effective means of sampling for gases and vapors and are ideal for personnel and area monitoring.
Supplier: Morphix Technologies 38000710
Description
- Built-in scale for direct reading of exposure dose features six dose ranges that change color upon exposure
- Color change and exposure measurement are easy to read, providing fast, accurate, onsite results
- Badge's patented design minimizes the effects of temperature, humidity, face velocity, and interferences
Order Info
Attaches to user's collar with badge clip (19-087-526, sold separately–contact your Fisher Safety Customer Service Representative for ordering information). Available 10 badges per pack.
Specifications
The ChromAir™ Formaldehyde badge may be used for personnel or area monitoring for exposure times ranging from 15 min. to 2 days | |
2 Days | |
61°F to 95°F | |
0.02 lb. | |
Formaldehyde Sampling Badge |
Formaldehyde | |
0.3 to 12 ppm.Hour | |
16°C to 35°C | |
0.01 kg |