Eisco™ Mortar and Pestle Set
Features a flat bottom, unglazed grinding surfaces, and spout for easy pouring. Eisco™ Mortar and Pestle Set is made from lab quality porcelain and designed for efficient and uniform grinding of various materials.
$8.62 - $108.74
Specifications
Product Type Mortar and Pestle
Material Porcelain
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Inner Price Quantity  
S23641
View Documents
Eisco™
CH0540B
148 mL 105 mm
Each for $16.18
S89316
View Documents
Eisco™
CH0540A
60 mL 80 mm
Each for $8.62
S23642
View Documents
Eisco™
CH0540C
275 mL 140 mm
Each for $17.97
S23643
View Documents
Eisco™
CH0540D
550 mL 153 mm
Each for $24.59
S39827
View Documents
Eisco™
CH0540E
900 mL 190 mm
Each for $44.82
S39828
View Documents
Eisco™
CH0540F
1500 mL 210 mm
Each for $70.41
S39829
View Documents
Eisco™
CH0540G
2000 mL 238 mm
Each for $108.74
Description
Description
  • Made from lab quality porcelain
  • Outside of mortar and handle of pestle is glazed
  • Inner bowl and pestle end are not glazed
  • Spout for easing pouring
  • Flat bottom for stability
Specifications
Specifications
Mortar and Pestle
Porcelain
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'