MP Biomedicals™ Aprotinin, From Bovine Lung
From Bovine Lung
Supplier:  MP Biomedicals™ 0219115883
 View more versions of this product
Catalog No. ICN19115883
Explore available promotions
Description
Description
Aprotinin, [9087-70-1]
  • C284H432N84O79S7
  • Lyophilized white powder
Specifications
Specifications
Inhibition Assay
Aprotinin from Bovine Lung
250 mg
4°C
6511.23
5 to 7
Bovine Lung
Lyophilized