missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Aprotinin, From Bovine Lung

From Bovine Lung

$97.92 - $3333.07

Specifications

For Use With (Application) Inhibition Assay
Molecular Weight (g/mol) 6511.23
Name Aprotinin from Bovine Lung
pH Range 5 to 7
Source Bovine Lung
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19115801
View Documents
MP Biomedicals™
0219115801
1 mg
Each for $97.92
 
ICN19115805
View Documents
MP Biomedicals™
0219115805
5 mg
Each for $146.44
 
ICN19115810
View Documents
MP Biomedicals™
0219115810
10 mg
Each for $207.74
 
ICN19115825
View Documents
MP Biomedicals™
0219115825
25 mg
Each for $385.96
 
ICN19115880
View Documents
MP Biomedicals™
0219115880
100 mg
Each for $1,249.96
 
ICN19115883
View Documents
MP Biomedicals™
0219115883
250 mg
Each for $3,333.07
 
Description

Description

Aprotinin, [9087-70-1]

  • C284H432N84O79S7
  • Lyophilized white powder

Specifications

Specifications

Inhibition Assay
Aprotinin from Bovine Lung
Bovine Lung
Lyophilized
6511.23
5 to 7
4°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'