MP Biomedicals™ Casein
Low Trace Metals
Supplier:  MP Biomedicals™ 0296012805
 View more versions of this product
Catalog No. ICN96012805
Explore available promotions
Description
Description
Precipitated from bovine milk by hydrochloric acid rather than lactic acid to remove most of the trace metals usually present in casein. Used in different animal diets.
Specifications
Specifications
Animal Diet
Powder
2.26 kg
15°C to 30°C
Protein 95%, Moisture 5%, Fat 1.5%, Lactose 0.2%, Ash 2.1%, pH (10% Solution) 4.6 to 5.1, Free Acids 0.2%
Used in different animal diet, blocking agen in immunochemistry
Rat