missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Casein

Low Trace Metals

$295.78 - $904.14

Specifications

Product Type Animal Diet
Components Protein 95%, Moisture 5%, Fat 1.5%, Lactose 0.2%, Ash 2.1%, pH (10% Solution) 4.6 to 5.1, Free Acids 0.2%
Format Powder
For Use With (Application) Used in different animal diet, blocking agen in immunochemistry
Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN96012805
View Documents
MP Biomedicals™
0296012805
2.26 kg
Each for $295.78
 
ICN96012825
View Documents
MP Biomedicals™
0296012825
11.33 kg
Each for $904.14
 
Description

Description

Precipitated from bovine milk by hydrochloric acid rather than lactic acid to remove most of the trace metals usually present in casein. Used in different animal diets.

Specifications

Specifications

Animal Diet
Powder
Rat
Protein 95%, Moisture 5%, Fat 1.5%, Lactose 0.2%, Ash 2.1%, pH (10% Solution) 4.6 to 5.1, Free Acids 0.2%
Used in different animal diet, blocking agen in immunochemistry
15°C to 30°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'