missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Dextrose

Monohydrate

Supplier:  MP Biomedicals™ 0290559405

 View more versions of this product

Catalog No. ICN90559405


Explore available promotions

Description

Description

Dextrose monohydrate is a common natural sugar involved in processes such as energy production, glycosylation and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.

Formula: C6H12O6•H2O

Formula Weight: 198.18

CAS No.: 14431-43-7

Specifications

Specifications

Animal Diet
Used as a supplement for cell culture and in numerous cellular processes
15°C to 30°C
Powder
2.26 kg
Videos
SDS
Documents

Documents

Promotions

Promotions