missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
MP Biomedicals™ Dextrose
Monohydrate
$90.22 - $252.19
Spécifications
Product Type | Animal Diet |
---|---|
Format | Powder |
For Use With (Application) | Used as a supplement for cell culture and in numerous cellular processes |
Storage Requirements | 15°C to 30°C |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
ICN90559405
|
MP Biomedicals™
0290559405 |
2.26 kg |
chaque for $90.22
|
|
|||||
ICN90559425
|
MP Biomedicals™
0290559425 |
11.33 kg |
chaque for $252.19
|
|
|||||
Description
Dextrose monohydrate is a common natural sugar involved in processes such as energy production, glycosylation and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.
Formula: C6H12O6•H2O
Formula Weight: 198.18
CAS No.: 14431-43-7
Spécifications
Animal Diet | |
Powder | |
Used as a supplement for cell culture and in numerous cellular processes | |
15°C to 30°C |