MP Biomedicals™ Gentamicin Sulfate
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Supplier:  MP Biomedicals™ 0219453005
 View more versions of this product
Catalog No. ICN19453005
Explore available promotions
Description
Description
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
General antimicrobial effective against gram positive bacteria, gram negative bacteria, and mycoplasma.
Specifications
Specifications
4°C
Antibiotic
3.5 to 5.5
Powder
Non-sterile
5 g