MP Biomedicals™ Glucose Oxidase
From Aspergillus niger
Supplier:  MP Biomedicals™ 0219519650
 View more versions of this product
Catalog No. ICN19519650
Explore available promotions
Description
Description
Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.
Specifications
Specifications
>100 U/mg
4 to 7
-20°C
50 kU
≈160000
Lyophilized Powder
Yellow
Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Aspergillus niger
Glucose Oxidase
Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose