MP Biomedicals™ Human Albumin
From human plasma
Supplier: MP Biomedicals™ 0219134901
Description
Negative for HBsAg and HIV antibodies as tested. Purity: >97%. Salt-free, lyophilized, homogeneous powder. MW: 65kDa.
Specifications
ELISA, Molecular Biology, Western Blot | |
Human Albumin | |
Human | |
Lyophilized |
65 | |
1 g | |
4°C |