missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Human Albumin
From human plasma
$577.29 - $2305.96
Specifications
For Use With (Application) | ELISA, Molecular Biology, Western Blot |
---|---|
Molecular Weight (g/mol) | 65 |
Name | Human Albumin |
Source | Human |
Storage Requirements | 4°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19134901
|
MP Biomedicals™
0219134901 |
1g |
Each for $577.29
|
|
|||||
ICN19134905
|
MP Biomedicals™
0219134905 |
5g |
Each for $2,305.96
|
|
|||||
Description
Negative for HBsAg and HIV antibodies as tested. Purity: >97%. Salt-free, lyophilized, homogeneous powder. MW: 65kDa.
Specifications
ELISA, Molecular Biology, Western Blot | |
Human Albumin | |
4°C |
65 | |
Human | |
Lyophilized |