missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ TeenPrep™ Lysing Matrix D Tubes
TeenPrep™ Lysing Matrix D Tubes contain 1.4 mm zirconium-silicate spheres and are used to grind soft animal and plant tissues, cultured cells and insects/worms.
$71.06 - $3103.44
Specifications
Bead Type | 1.4 mm Zirconium-silicate Spheres |
---|---|
Sample Type | Animal Tissues, Plants, Cultured Cells, Insects, Worms |
DNase | Dnase-free |
For Use With (Application) | Nucleic Acid Isolation and Purification, Homogenization |
Matrix | Lysing Matrix D |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
MP116933050
|
MP Biomedicals™
116933050 |
50 x 15 mL |
Each for $684.60
|
|
|||||
MP116933005
|
MP Biomedicals™
116933005 |
5 x 15 mL |
Each for $71.06
|
|
|||||
MP116933025
|
MP Biomedicals™
116933025 |
25 x 15 mL |
Each for $455.46
|
|
|||||
MP116973025
|
MP Biomedicals™
116973025 |
25 x 4.5 mL Tubes |
Each for $305.29
|
|
|||||
MP116983001
|
MP Biomedicals™
116983001 |
1 x 96-Well Plate |
Each for $463.19
|
|
|||||
MP116913050
|
MP Biomedicals™
116913050 |
50 x 2 mL Tubes |
Each for $254.38
|
|
|||||
MP116953010
|
MP Biomedicals™
116953010 |
10 x 50 mL |
Each for $152.52
|
|
|||||
MP116973050
|
MP Biomedicals™
116973050 |
50 x 4.5 mL Tubes |
Each for $557.31
|
|
|||||
MP116983010
|
MP Biomedicals™
116983010 |
10 x 96-Well Plate |
Each for $3,103.44
|
|
|||||
MP116913100
|
MP Biomedicals™
116913100 |
100 x 2 mL Tubes |
Each for $498.50
|
|
|||||
MP116953050
|
MP Biomedicals™
116953050 |
50 x 50 mL |
Each for $990.13
|
|
|||||
MP116973100
|
MP Biomedicals™
116973100 |
100 x 4.5 mL Tubes |
Each for $959.21
|
|
|||||
MP116913500
|
MP Biomedicals™
116913500 |
500 x 2 mL Tubes |
Each for $2,545.90
|
|
|||||
MP116953100
|
MP Biomedicals™
116953100 |
100 x 50 mL |
Each for $1,377.79
|
|
|||||
Description
TeenPrep™ Lysing Matrix D, tubes can be used with FastPrep instruments or similar bead beater. The grinding matrix is chemically inert and will not bind nucleic acids.
- Tube size: 15 mL
- Sample Types: Soft tissues such as brain, liver, kidney, lung and spleen; plant tissues such as leaves, roots and fruits; cultured cells; whole insects; nematodes
- Matrix Composition: 1.4 mm zirconium-silicate spheres
Specifications
1.4 mm Zirconium-silicate Spheres | |
Dnase-free | |
Lysing Matrix D |
Animal Tissues, Plants, Cultured Cells, Insects, Worms | |
Nucleic Acid Isolation and Purification, Homogenization | |
Rnase-free |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.