missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Lysozyme from Chicken Egg White, 20,000U/mg

Hydrolyzes b-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine which occur in mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus

Supplier:  MP Biomedicals™ 0210083125

 View more versions of this product

Catalog No. ICN10083125


Explore available promotions

Description

Description

  • Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
  • CAS Number: 9001-63-2
  • EC Number: 232-620-4
  • MDL Number: MFCD00131557
  • Format: 3X crystallized, salt-Free, albumin-Free, lyophilized
  • Specific Activity: ≤20,000U/mg solid
  • Extinction Coefficient (E1%): 26.4 (280 nm)
  • Isoelectric point (pI): pH 11.0
  • pH: 3.6

Used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Also acts as germinative agent of bacterial spores. In humans, lysozyme may mediate anti-tumor function of macrophages. Cartilage lysozyme has a role in cartilage calcification.

Specifications

Specifications

3X
11
0°C
25 g
14388
Crystallized, Lyophilized
White
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen
Chicken egg white
Lysozyme
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes
Videos
SDS
Documents

Documents

Promotions

Promotions