missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Lysozyme from Chicken Egg White, 20,000U/mg
Hydrolyzes b-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine which occur in mucopeptide cell wall structure of certain microorganisms, such as Micrococcus lysodeikticus
$80.25 - $2256.68
Specifications
Concentration | 3X |
---|---|
Color | White |
pH | 11 |
For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
Content And Storage | 0°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10083101
|
MP Biomedicals™
0210083101 |
1 g |
Each for $80.25
|
|
|||||
ICN10083105
|
MP Biomedicals™
0210083105 |
5 g |
Each for $229.69
|
|
|||||
ICN10083110
|
MP Biomedicals™
0210083110 |
10 g |
Each for $353.71
|
|
|||||
ICN10083125
|
MP Biomedicals™
0210083125 |
25 g |
Each for $711.29
|
|
|||||
ICN10083180
|
MP Biomedicals™
0210083180 |
100 g |
Each for $2,256.68
|
|
|||||
Description
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS Number: 9001-63-2
- EC Number: 232-620-4
- MDL Number: MFCD00131557
- Format: 3X crystallized, salt-Free, albumin-Free, lyophilized
- Specific Activity: ≤20,000U/mg solid
- Extinction Coefficient (E1%): 26.4 (280 nm)
- Isoelectric point (pI): pH 11.0
- pH: 3.6
Used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Also acts as germinative agent of bacterial spores. In humans, lysozyme may mediate anti-tumor function of macrophages. Cartilage lysozyme has a role in cartilage calcification.
Specifications
3X | |
11 | |
0°C | |
Lysozyme | |
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
White | |
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen | |
Chicken egg white | |
14388 | |
Crystallized, Lyophilized |