missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Ribonuclease A
From bovine pancreas
Supplier: MP Biomedicals™ 0219398010
Description
Chromatographically purified. Salt-free and protease-free. Activity: ≥70 Kunitz units/mg. Unit Definition: One unit causes the hydrolysis of RNA at a rate such that the velocity constant (k) equals 1 at 25°C and pH 5.0. Lyophilized powder.
Specifications
≥70 kU/mg | |
>70% | |
purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples | |
Bovine pancreas | |
Ribonuclease A | |
Heavy Metal Ions, DNA |
White | |
7 to 7.5 | |
0°C | |
10 mg | |
13700 | |
Lyophilized Powder |