missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Ribonuclease A

From bovine pancreas

$99.49 - $3115.49

Specifications

Concentration ≥70 kU/mg
Color White
Purity >70%
pH 7 to 7.5
For Use With (Application) purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19398010
View Documents
MP Biomedicals™
0219398010
10 mg
Each for $99.49
 
ICN19398083
View Documents
MP Biomedicals™
0219398083
250 mg
Each for $1,086.36
 
ICN19398050
View Documents
MP Biomedicals™
0219398050
50 mg
Each for $297.11
 
ICN19398091
View Documents
MP Biomedicals™
0219398091
1 g
Each for $3,115.49
 
ICN19398090
View Documents
MP Biomedicals™
0219398090
500 mg
Each for $1,842.97
 
Description

Description

Chromatographically purified. Salt-free and protease-free. Activity: ≥70 Kunitz units/mg. Unit Definition: One unit causes the hydrolysis of RNA at a rate such that the velocity constant (k) equals 1 at 25°C and pH 5.0. Lyophilized powder.

Specifications

Specifications

≥70 kU/mg
>70%
purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
Bovine pancreas
13700
Lyophilized Powder
White
7 to 7.5
0°C
Ribonuclease A
Heavy Metal Ions, DNA
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'