missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Ribonuclease A

From bovine pancreas

Supplier:  MP Biomedicals™ 0219398050

 View more versions of this product

Catalog No. ICN19398050



Description

Description

Chromatographically purified. Salt-free and protease-free. Activity: ≥70 Kunitz units/mg. Unit Definition: One unit causes the hydrolysis of RNA at a rate such that the velocity constant (k) equals 1 at 25°C and pH 5.0. Lyophilized powder.

Specifications

Specifications

≥70 kU/mg
>70%
purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
Bovine pancreas
Ribonuclease A
Heavy Metal Ions, DNA
White
7 to 7.5
0°C
50 mg
13700
Lyophilized Powder
SDS
Documents

Documents

Promotions

Promotions