missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ MQuant™ Peroxide Test Strips

To check for presence or absence of residues of Peroxide

Supplier:  MilliporeSigma™ 1100110001

 View more versions of this product

Catalog No. M1100110001


Explore available promotions

Includes: Tube containing 100 strips

Description

Description

  • Small and easy to handle
  • Flexible and universal application
  • Enzymatic reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10011.0001
Color-scale graduation: 0.5 - 2 - 5 - 10 - 25mg/L H2O2

1.10081.0001
Color-scale graduation: 1 - 3 - 10 - 30 - 100mg/L H2O2

1.10337.0001
Color-scale graduation: 100 - 200 - 400 - 600 - 800 - 1000mg/L H2O2

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

Specifications

For testing residual concentrations of disinfectants on equipment or the product itself
0.5-2-5-10-25
100 Strips/Pk.
2°C to 8°C
Peroxide Test Strips
Colorimeter
100
0.5 to 25mg/L H2O2
Enzymatic Reaction
Videos
SDS
Documents

Documents

Promotions

Promotions