missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ MQuant™ Peroxide Test Strips

To check for presence or absence of residues of Peroxide

$142.53 - $320.45

Specifications

For Use With (Application) For testing residual concentrations of disinfectants on equipment or the product itself
For Use With (Equipment) Colorimeter
Storage Requirements 2°C to 8°C
Test Method Enzymatic Reaction
Type Peroxide Test Strips
View More Specs

Products 4
Catalog Number Mfr. No. Range Graduations Quantity Price Quantity  
Catalog Number Mfr. No. Range Graduations Quantity Price Quantity  
M1100810001
View Documents
MilliporeSigma™
1100810001
10 to 100mg/L H2O2 1-3-10-30-100 100 Strips/Pk.
Each for $200.08
 
M1100110001
View Documents
MilliporeSigma™
1100110001
0.5 to 25mg/L H2O2 0.5-2-5-10-25 100 Strips/Pk.
Each for $203.66
 
M1103370001
View Documents
MilliporeSigma™
1103370001
100 to 1000mg/L H2O2 100 - 200 - 400 - 600 - 800 - 1000 100 Strips/Pk.
Each for $320.45
 
M1100110002
View Documents
MilliporeSigma™
1100110002
0.5 to 25mg/L H2O2 0.5 - 2 - 5 - 10 - 25 25 strips/Pk.
Each for $142.53
 
Description

Description

  • Small and easy to handle
  • Flexible and universal application
  • Enzymatic reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10011.0001
Color-scale graduation: 0.5 - 2 - 5 - 10 - 25mg/L H2O2

1.10081.0001
Color-scale graduation: 1 - 3 - 10 - 30 - 100mg/L H2O2

1.10337.0001
Color-scale graduation: 100 - 200 - 400 - 600 - 800 - 1000mg/L H2O2

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

Specifications

For testing residual concentrations of disinfectants on equipment or the product itself
2°C to 8°C
Peroxide Test Strips
Colorimeter
Enzymatic Reaction
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'